Learn More
Invitrogen™ Human S100 A7 ELISA Kit
Human S100 A7 quantitates human S100 A7 in serum, plasma, supernatant.
Brand: Invitrogen™ EH400RB
564.16 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 2.50000kg
Description
Human S100 A7 quantitates human S100 A7 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human S100 A7.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known.Specifications
protein S100-A7; PSOR1; Psoriasin; psoriasin 1; S100 calcium binding protein A7; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100A7; S100A7C | |
24 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
S100A7 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
24 to 6,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P31151 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL | |
S100A7 | |
4 hr 45 min |