missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WSB1 Partial ORF (NP_056441, 101 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_056441 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26118 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035385
|
Abnova™
H00026118-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025385
|
Abnova™
H00026118-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the WD-protein subfamily. This protein shares a high sequence identity to mouse and chick proteins. It contains several WD-repeats spanning most of the protein and an SOCS box in the C-terminus. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: EHIIDCGDIVWSLAFGSSVPEKQSRCVNIEWHRFRFGQDQLLLATGLNNGRIKIWDVYTGKLLLNLVDHTEVVRDLTFAPDGSLILVSASRDKTLRVWDLSpecifications
NP_056441 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SWIP1/WSB-1 | |
WSB1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26118 | |
WSB1 (Human) Recombinant Protein (Q01) | |
EHIIDCGDIVWSLAFGSSVPEKQSRCVNIEWHRFRFGQDQLLLATGLNNGRIKIWDVYTGKLLLNLVDHTEVVRDLTFAPDGSLILVSASRDKTLRVWDL | |
RUO | |
WSB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |