Learn More
Cytiva HyClone™ Antibiotic/Antimycotic Solution (Pen/Strep/Fungiezone) solution
Filter sterilized. Cytiva HyClone™ Antibiotic Antimycotic (Pen/Strep/Fungiezone) Solution controls or destroys destructive microorganisms such as bacteria, fungi and yeast.
Brand: Cytiva SV30079.01
51.46 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : 83
Additional Details : Weight : 0.00900kg
Description
- Contains 10,000 units of penicillin, 10,000μg of streptomycin, and 25μg of Amphotericin B per mL
- Wide spectrum of activity against gram negative and gram positive bacteria as well as activity against fungi and yeast
- Commonly used as an antifungal, anti-yeast, anti-mold or antibacterial
- Common working dose is 10mL/L of 100X solution
Specifications
Store frozen | |
Broad spectrum antibacterial and antifungal | |
Liquid | |
100 mL |
Antibiotic Antimycotic Solution (Pen/Strep/Fungizone), 100X | |
100 X | |
0.2 μm Sterile-filtered |
Safety and Handling
- HyClone Antibiotic/Antimycotic Solution
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : streptomycin sulphate