missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)
Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.
Brand: Cytiva SH30084.03IH
Code : 84
Additional Details : Weight : 0.50000kg
Description
- Filtered through 3 sequential 0.1 μm pore size filters
Specifications
AQIS | |
Australia | |
500 mL | |
Sequential Triple-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
HyClone™ |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Fetal Bovine Serum | |
Sterile | |
Heat Inactivated | |
Yes | |
USDA Inspected |