missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Clone I Serum
Brand: Cytiva SH30080.03HI
Code : A0
Additional Details : Weight : 0.01000kg
Specifications
500mL | |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG | |
Yes | |
≤ 20 mg/dL | |
U.S. | |
Heat Inactivated | |
Store at –10°C or below | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
FetalClone I |
Sterile | |
FetalClone I Serum (U.S.) Heat Inactivated | |
≤ 10 EU/mL | |
Typically less than 2 μg/mL | |
U.S. | |
No | |
1mL | |
Filtered using sequential 100 nm pore-size rated filters. | |
USDA Inspected | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |