missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ II Serum (U.S.)
Developed for growth of CHO cells and derivatives, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ II Serum, a fetal bovine serum (FBS) alternative, containing additional growth factors and supplements than basic formulations.
91.70 € - 382.00 €
Specifications
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
---|---|
Description | FetalClone II Serum (U.S.) |
Country of Origin | United States |
Product Type | FBS Replacement Serum |
Sterility | Sterile |
Product Code | Brand | Description | Quantity | Processing | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Processing | Price | Quantity & Availability | |||
10041492
|
Cytiva
SH30066.02 |
FetalClone II Serum (U.S.) | 100 mL | None |
91.70 €
100mL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10326762
|
Cytiva
SH30066.03 |
FetalClone II Serum (U.S.) | 500 mL | None |
382.00 €
500mL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Economical alternative to FBS
- Performance comparable with FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Specifications
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
Sterile | |
None | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
No |
FetalClone II Serum (U.S.) | |
FBS Replacement Serum | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Typically less than 2 μg/mL | |
USDA Inspected |