missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum, U.S. Origin
A high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. Demonstrates excellent performance with a wide variety of cell types.
Brand: Cytiva SH30087.03IH
Code : 84
Additional Details : Weight : 0.50000kg
Description
- Age of animal at time of collection ranges from 16 to 24 weeks
- Supplemented with iron and growth-promoting factors
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
- Sourced in the USA with complete traceability back to original source
Specifications
Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Iron, TIBC, Percent Saturation, Glucose, and IgG | |
Store at or below -10°C | |
United States | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, and Reovirus |
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities | |
16 to 22 weeks | |
500 mL | |
Gamma Irradiated, Heat Inactivated | |
Growing a Broad Spectrum of Cell Types |