Learn More
HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™
Brand: Honeywell Fluka 34840-50ML
Code : NEW
Additional Details : Weight : 0.05320kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Specifications
14°C (57.2°F) | |
HYDRANAL™-Coulomat CG | |
UN1230 |
10 x 5 mL Ampules | |
14426-21-2,67-56-1,134227-25-1,7446-09-5 |
Safety and Handling
- HYDRANAL-Coulomat CG
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, diethanolamine hydrochloride,Ethanol, 2,2'-iminobis-, hydriodide, sulphur dioxide