Learn More
HYDRANAL™ - Working Medium K, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™
Brand: Honeywell Fluka 34817-1L
Code : NEW
Additional Details : Weight : 1.35000kg Transport : UN number : 3382 Chem class : 6.1 Pack group : I
Specifications
>100°C (>212°F) | |
HYDRANAL™-Working Medium K | |
67-66-3,107-07-3 | |
UN3382 |
1 L | |
1.34g/mL | |
MFCD00147202 |
Safety and Handling
- HYDRANAL-Working Medium K
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trichloromethane; chloroform, 2-chloroethanol; ethylene chlorohydrin