missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+

Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.

Brand:  Lovibond™ 462910

Code : NEW

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15321715

  • 72.90 € / 30g
Estimated Shipment: 25-04-2024
to see stock.
Add to basket

Description

Description

Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.

  • Highest quality production standards
  • Consistent and reliable reagents are provided in liquid and tablet formats
  • Maximum accuracy
Specifications

Specifications

Hydrazine Test Powder
Water analysis
30 g
Powder
Colorimeter and spectrophotometer
Comparator 2000+ replacement reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers