missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 37%, Extra Pure, d=1.18, SLR, Fisher Chemical™
18.81 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
25.21 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "23271" to get your promotional price.
25.21 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Description
ClH, CAS Number-7647-01-0, hydrogen chloride hcl, muriatic acid, hydrochloric acid, chlorwasserstoff, hydrogen chloride, chlorohydric acid, chloorwaterstof, acide chlorhydrique, spirits of salt, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Specifications
pH | 1 |
Color | Colorless |
Melting Point | -35°C |
Boiling Point | 57°C |
Packaging | HDPE plastic bottle |
Vapor Pressure | 125mbar at 20°C |
Quantity | 1 L |
Viscosity | 1.8 mPaS at 15°C |
Physical Form | Liquid |
Formula Weight | 36.46g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'