missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
116.58 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
19 L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle
Poly Pail
Specifications
Packaging | Narrow Mouth Polyethylene Bottle |
Specific Gravity | 1L = 1.08kg |
Quantity | 1 L |
IUPAC Name | hydrogen chloride |
Formula Weight | 36.46 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Hydrochloric Acid, 5N Volumetric Solution |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'