missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Multi-Compendial, N.F., J.T.Baker™

Brand:  JT Baker 9544-05

Code : NEW

Additional Details : CAS Number : 7647-01-0 Weight : 2.95000kg Transport : UN number : 1789 Chem class : 8 Pack group : II

Packaging Glass Bottle, Single ship
Quantity 2.5L

 View more versions of this product

Product Code. 15394468

  • 142.00 € / 2.50 L


Call for latest local availability
01 885 5854
Add to basket

Specifications

Chemical Identifiers

7647-01-0
36.458
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane

Specifications

Hydrochloric Acid
7647-01-0
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.458
CHEBI:17883
Multi-Compendial/NF
1L = 1.18kg
2.5L
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46
Glass Bottle, Single ship
Videos