Learn More
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, acide chlorhydrique, hydrogen chloride hcl, hydrogen chloride, chlorohydric acid, chlorwasserstoff, hydrochloric acid, spirits of salt, muriatic acid, 1L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Brand: Fisher Chemical J/4320/15
32.15 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : 23
Additional Details : CAS Number : 7647-01-0 Weight : 1.02000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, acide chlorhydrique, hydrogen chloride hcl, hydrogen chloride, chlorohydric acid, chlorwasserstoff, hydrochloric acid, spirits of salt, muriatic acid, 1L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BPSpecifications
Colorless | |
100°C | |
1 L | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
7647-01-0 | |
HDPE plastic bottle | |
ClH | |
MFCD00011324 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46g/mol |