Learn More
Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, acide chlorhydrique, chlorohydric acid, hydrochloric acid, chloorwaterstof, hydrogen chloride hcl, anhydrous hydrochloric acid, hydrogen chloride, muriatic acid, spirits of salt, chlorwasserstoff, 1L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Brand: Fisher Chemical A508-P1
148.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 25
Additional Details : CAS Number : 7647-01-0 Weight : 1.18000kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
ClH, CAS Number-7647-01-0, acide chlorhydrique, chlorohydric acid, hydrochloric acid, chloorwaterstof, hydrogen chloride hcl, anhydrous hydrochloric acid, hydrogen chloride, muriatic acid, spirits of salt, chlorwasserstoff, 1L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol
PolybottleSpecifications
1 | |
-35°C | |
57°C | |
1 L | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46g/mol |
Colorless | |
7647-01-0 | |
125mbar at 20°C | |
1.8 mPaS at 15°C | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Safety and Handling
- Hydrochloric acid, 34-37%
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid