missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution, Contains inhibitor, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Color | Colorless |
UN Number | UN2014 |
Beilstein | 3587191 |
Density | 1.11g/mL at 25°C |
Solubility Information | Completely miscible in water |
Formula Weight | 34.01g/mol |
Odor | Stinging |
Packaging | Poly bottle |
Vapor Pressure | 23.3mmHg (30°C) |
pH | 2.0 to 4.0 (at 20°C) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'