missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyacetone, Technical, Thermo Scientific Chemicals

Catalog Number p-3771115
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
116-09-6
C3H6O2
74.08
MFCD00004669
XLSMFKSTNGKWQX-UHFFFAOYSA-N
hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol
8299
CHEBI:27957
1-hydroxypropan-2-one
CC(=O)CO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
116-09-6
C3H6O2
74.08
MFCD00004669
XLSMFKSTNGKWQX-UHFFFAOYSA-N
hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol
8299
CHEBI:27957
1-hydroxypropan-2-one
CC(=O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -17.0°C
Density 1.0800g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 56°C
Packaging Glass bottle
Linear Formula CH3COCH2OH
Refractive Index 1.4315
Quantity 100 g
UN Number 1224
Beilstein 01, 821
Merck Index 15, 64
Specific Gravity 1.08
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Viscosity 1.593 mPa.s (48°C)
Formula Weight 74.08
Grade Technical
Chemical Name or Material Hydroxyacetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'