missing translation for 'onlineSavingsMsg'
Learn More

Hydroxymethylpolystyrene resin, 1% crossl. with DVB, 1.0 to 1.2mmol/g, 100 to 200 mesh

Brand:  Acros Organics 349510050

Code : 29

Additional Details : CAS Number : 66072-40-0 Weight : 0.05500kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10002061

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.


Description

Description

For solid support synthesis
Specifications

Chemical Identifiers

Hydroxymethylpolystyrene resin
66072-40-0

Specifications

Hydroxymethylpolystyrene resin
66072-40-0
C7H8O
C1=CC=C(C=C1)CO
108.14
CHEBI:17987
5g
Glass bottle
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
Videos
SDS