Learn More
Ansell HyFlex™ 11-953 Rough Textured Nitrile-Coated Gloves
Rough texture on the palm protects against oil, while also enhancing grip capabilities in food-safe environments.
Brand: Ansell 11953080
Additional Details : Weight : 7.20000kg
Description
- Rough nitrile palm coating
- Protects against oil
- Enhances grip in food environments
Specifications
EN 420:2003 + A1:2009, REACH Compliant | |
Gray | |
Polyamide | |
4121A | |
Knit | |
13 |
Nitrile | |
Specific | |
8 | |
Assembly, Maintenance, Metal Fabrication, and General Handling | |
11-953 |