Learn More
Plas Labs™ Hypalon Gloves
Click to view available options
Size:
7
Description
- White one-piece gauntlet style glovebox gloves
- Constructed from Hypalon™ chlorosulfonated polyethylene (CSPE) synthetic rubber
- Resistant to abrasion, oxidising chemicals ozone and UV light
- Ambidextrous fit
- Sold in pairs
- Range of hand sizes and port diameters
Specifications
Material | CSM |
Product Type | Gloves |
Size | 7 |
Thickness (Metric) | 0.15 mm |