Learn More
Thermo Scientific™ HyperSep™ Retain CX SPE 96-Well Plates and Individual Wells
Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain™ CX SPE 96-Well Plates and Individual Wells.
Brand: Thermo Scientific™ 60303-308
Code : 16
Additional Details : Weight : 0.30000kg
Description
Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain™ CX SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with sulfonic acid functional groups to give superior recoveries for basic analytes. Used in SPE sample preparation, these cation exchange well plates and individual wells can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Widely used in forensic and clinical analysis of a wide range of drugs of abuse, including basic and neutral drugs. Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.- High, consistent recoveries
- Ideal for large samples
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- pH stable 0 to 14
- Available in a wide range of chemistries and bed weights
Specifications
96-well plate; Bed weight 60 mg | |
30 to 50 μm | |
HyperSep Retain CX |
Analysis of Drugs of Abuse | |
HyperSep | |
96-well Plate |