missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ UniGuard Direct-Connect Online SPE Columns

Product Code. 10005984 Shop All Thermo Scientific Products
Click to view available options
Sorbent:
HyperSep Hypercarb
HyperSep Quaternary Amine-Modified Polystyrene DVB
HyperSep Sulfonic Acid-Modified Polystyrene DVB
HyperSep Urea-Modified Polystyrene DVB
Diameter (Metric) Inner:
2.1 mm
3.0 mm
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 10005984

Brand: Thermo Scientific™ 60311401

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Clean-up and concentrate target analytes in minutes with these direct-connection SPE cartridges, offering quick, easy connection via a reusable drop-in cartridge holder.

Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:

  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds

Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Column offer quick, easy connection by means of a reusable drop-in guard cartridge holder.

HyperSep UNIGUARD Direct-Connect Online SPE Cartridges attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries:

HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes

HyperSep Retain-CX — for retention of basic and nonpolar analytes

HyperSep Retain-AX — for retention of acidic and nonpolar analytes

HyperSep™ Hypercarb™ — for retention of extremely polar analytes

TRUSTED_SUSTAINABILITY

Specifications

Description HyperSep Retain-AX; 2.1 mm ID
For Use With (Application) Retention of Acidic Analytes
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Quaternary Amine-Modified Polystyrene DVB
Type Online Solid Phase Extraction Cartridge
Quantity Pack of 4
Diameter (Metric) Inner 2.1 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.