missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ ODS-2 C18 Columns

Use as a general purpose C18 phase for acidic and neutral analytes separated under difficult operating conditions.

Brand:  Thermo Scientific™ 31603-104030

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10007514

  • 520.00 € / Each


Estimated Shipment date
from Supplier 25-03-2020
Add to basket

Description

Description

  • Based on Type A silica
  • Excellent reproducibility
  • Selectivity over wide range of applications
  • Manufactured under ISO 9001:2000 quality guidelines

Ordering Information:

Other column dimensions are available. Please call your Customer Service Representative for more information.

Specifications

Specifications

11%, 11%, 11%, 11%, 11%
Reversed Phase
Yes
100 mm
Silica, Spherical Fully Porous Type A
2 to 8, 2 to 8, 2 to 8, 2 to 8, 2 to 8
80 Å
60°C, 60°C, 60°C, 60°C, 60°C
C18, C18, C18, C18, C18
Analytical Column, Analytical Column, Analytical Column, Analytical Column, Analytical Column
4 mm
HPLC, HPLC, HPLC, HPLC, HPLC
5800 psi (400 bar), 5800 psi (400 bar), 5800 psi (400 bar), 5800 psi (400 bar), 5800 psi (400 bar)
3 µm
Reversed Phase, Reversed Phase, Reversed Phase, Reversed Phase
220 m²/g, 220 m²/g, 220 m²/g, 220 m²/g, 220 m²/g
L1, L1, L1, L1, L1
Videos
Documents

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.