missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ I-Block™ Protein-Based Blocking Reagent

Highly purified Casein-based blocking reagent. Thermo Scientific I-Block Protein-Based Blocking Reagent provides superior blocking compared to both dried milk and BSA.

Brand:  Invitrogen™ T2015

Additional Details : Weight : 0.10000kg

Product Code. 12684305

  • 114.00 € / 30g
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Biotin-free
  • Tested in assays using Tropix substrates and alkaline phosphatase conjugates
  • Useful as blocking reagent in membrane-based and immunoassay applications
  • Suggested working
  • concentration is 0.2% (w/v) for detection of nucleic acids on neutral or positively-charged nylon membranes and for immunoassays and protein detection on membranes (nitrocellulose, PVDF, or neutral nylon)
  • Concentration of 3% is recommended for protein detection on positively-charged nylon membrane (such as TropilonPlus™ membrane)
Preparation
Prepared in either Tris- or phosphate-buffered saline buffer with heating (40° to 50°C).
 
Applications
Can be used with Western-Light™, Western-Light Plus™, Western-Star™, ELISA-Light™, Southern-Light™, and Southern-Star™ Detection Systems.
Specifications

Specifications

Reagent
I-Block™, NovaBright™
Blocking Buffers
Room Temperature
30 g
Western-Light™ Detection System, Western-Star™ Detection System, Southern-Star™ Detection System, ELISA-Light™ Detection System, Southern-Light™ Detection System, Western-Light Plus™ Detection System
Store the product at room temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.