Learn More
Brady™ IDXPERT™ MRO Safety and Facility Labels
IDXPERT™ MRO Safety and Facility Labels are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Brand:  Brady™ 142329
Code : NEW
Additional Details : Weight : 0.09980kg
 View more versions of this product
Product Code. 11831633
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • All-in-one cartridge includes ribbon and label supply
Specifications
Specifications
UL
Acrylic, Permanent
Gloss
Safety and Facility Label
General and industrial identification, General ID, General identification, Laboratory identification, Outside-of-panel markings, Panel identification, Voice/Data identification
50 Gardner Units
1 Ea.
-40°C to +120°C
0.102 mm
19 in.
1.5 in.
Powder Coated, Smooth, Textured/Rough
White, Green
Vinyl
Non-sterile
iDXPERT™, LabXpert™ Printer
1
-104 to 248°F
0.004 in.
9.14 m
38.10 mm
3.81 cm