missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ IEF Sample Buffer, 2X, pH 3-10 with lysine and arginine
38.10 € - 55.10 €
Specifications
Concentration | 2X |
---|---|
pH Range | 3 to 10 |
Product Type | Sample Loading Buffer |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15470237
|
Thermo Scientific™
J62241.AC |
25 mL |
38.10 €
25mL |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15480237
|
Thermo Scientific™
J62241.AD |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2X | |
3 to 10 | |
Sample Loading Buffer | |
Liquid |