missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) Mouse, Clone: rIG266, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) |
---|---|
Clone | rIG266 |
Concentration | 200μg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930218
|
EnquireBio
3500-MSM10-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920218
|
EnquireBio
3500-MSM10-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
IgG1 | |
Protein A/G purified from Bioreactor concentrate. | |
Cytoplasm, Cell Surface and Secreted | |
293T, Raji or hPBL cells. Tonsil or Spleen. |
rIG266 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
RUO | |
Purified human Ig Gamma Chain | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
75kDa |