missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Rat, PE, Clone: MP4-25D2, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB82-PE-100Tests
Additional Details : Weight : 0.01000kg
Specifications
IL-4 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
IL4 | |
Protein A/G Purified | |
RUO | |
3565 | |
IgG1 κ |
Flow Cytometry | |
MP4-25D2 | |
PE | |
NP_758858 | |
Rat | |
100 Tests | |
Primary | |
Store at 2-4°C. |