Learn More
Invitrogen™ IL-8 Canine ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ECCXCL8X5
Additional Details : Weight : 0.33820kg
Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Canine Interleukin 8 (Dog IL-8) ELISA quantitates Dog IL-8 in canine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Dog IL-8. Principle of the method The Canine IL-8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-8 (Interleukin-8) is a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8, secreted by endothelial cells and monocytes, which acts as a major chemoattractant for neutrophils and also T cells and basophils. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C.Specifications
P41324 | |
6 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
Colorimetric Microplate Reader | |
IL8 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
IL8 | |
1 hr. 20 min. |
6.14 pg/mL to 1500 pg/mL | |
6 pg/mL to 1500 pg/mL | |
Biotin | |
GCPR | |
This ELISA antibody pair detect canine IL-8. Other species not determined yet. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
403850 | |
<12% | |
Biotinylated Detection Antibody, Chromogen, SAV-HRP, Standard, Pre-coated 96 well plate, Adhesive Plate Covers, Assay Diluent concentrate, Stop Solution, Wash Buffer | |
5 x 96 Tests | |
Extracellular Matrix & Cell Adhesion | |
2°C to 8°C | |
Canine | |
4 hr. 45 min. |