missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-2-carboxaldehyde, 97%, Thermo Scientific™

Catalog Number p-7077389
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
118000-43-4
C8H6N2O
146.15
QKKKBOZQZHBENG-UHFFFAOYSA-N
imidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde, 2-formylpyridoimidazole, 2,3-dihydroimidazo 1,2-a pyridine-2-carbaldehyde, acmc-20ac55, imidazo 1,2-apyridine-2-carbaldehyde, imid-azo 1,2-a pyridine-2-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde 9ci
4961257
imidazo[1,2-a]pyridine-2-carbaldehyde
C1=CC2=NC(=CN2C=C1)C=O
This item is not returnable. View return policy
118000-43-4
C8H6N2O
146.15
QKKKBOZQZHBENG-UHFFFAOYSA-N
imidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde, 2-formylpyridoimidazole, 2,3-dihydroimidazo 1,2-a pyridine-2-carbaldehyde, acmc-20ac55, imidazo 1,2-apyridine-2-carbaldehyde, imid-azo 1,2-a pyridine-2-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde 9ci
4961257
imidazo[1,2-a]pyridine-2-carbaldehyde
C1=CC2=NC(=CN2C=C1)C=O
Melting Point 117.0°C to 119.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material Imidazo[1,2-a]pyridine-2-carboxaldehyde
compliance-icons
Product Identifier
  • Imidazo¢1,2-a!pyridine-2-carboxaldehyde, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'