missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[2,1-b][1,3]thiazole-6-carboxylic acid, ≥97%, Thermo Scientific™

Catalog Number p-3110995
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
53572-98-8
C6H4N2O2S
168.17
MFCD03419463
BAKIBSRDBASFAQ-UHFFFAOYSA-N
imidazo 2,1-b thiazole-6-carboxylic acid, imidazo 2,1-b 1,3 thiazole-6-carboxylic acid, imidazo 2,1-b 1,3-thiazoline-6-carboxylic acid
2759354
imidazo[2,1-b][1,3]thiazole-6-carboxylic acid
C1=CSC2=NC(=CN21)C(=O)O
This item is not returnable. View return policy
53572-98-8
C6H4N2O2S
168.17
MFCD03419463
BAKIBSRDBASFAQ-UHFFFAOYSA-N
imidazo 2,1-b thiazole-6-carboxylic acid, imidazo 2,1-b 1,3 thiazole-6-carboxylic acid, imidazo 2,1-b 1,3-thiazoline-6-carboxylic acid
2759354
imidazo[2,1-b][1,3]thiazole-6-carboxylic acid
C1=CSC2=NC(=CN21)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 168.18
Percent Purity 97%
Chemical Name or Material Imidazo[2, 1-b][1, 3]thiazole-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'