Learn More
Imidazole, Pure, Fisher Chemical™
Brand: Fisher Chemical I/0010/53
Code : 21
Additional Details : CAS Number : 288-32-4 Weight : 0.50000kg Transport : UN number : 3263 Chem class : 8 Pack group : III
Quantity | 500g |
---|
Chemical Identifiers
288-32-4 | |
68.079 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
795 | |
1H-imidazole |
C3H4N2 | |
5183 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
CHEBI:16069 | |
C1=CN=CN1 |
Specifications
Plastic powder jar | |
255°C | |
86°C | |
10.5 | |
C3H4N2 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
C1=CN=CN1 | |
68.079 | |
CHEBI:16069 | |
Solid |
0.003mbar at 20°C | |
White to Yellow | |
500g | |
288-32-4 | |
5183 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
1H-imidazole | |
795 | |
68.08g/mol |
Safety and Handling
- Imidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.