missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole Parmer In-Line Gas/Liquid Filters
Cole-Parmer In-Line Gas/Liquid Filters have a borosilicate glass filter membrane.
Brand: Cole Parmer 02909-30
Code : A0
Additional Details : Weight : 0.01000kg
Description
Cole-Parmer In-Line Gas/Liquid Filters have a borosilicate glass filter membrane.
- Filter gases or liquids with these microfiber filters
- All models have a borosilicate glass filter membrane
- Liquid efficiency ratings are 98% of retention of particles of noted size
- Gas efficiency ratings are percentage retention of 0.01 μm particles.
- Max differential pressure for all filters is 40 psi (60 psi for 0.01 μm units).
- Max temperature at 0 psi: nylon is 230°F; stainless steel and KYNAR PVDF are 250°F
Specifications
1/4 in. | |
5/Pk. | |
In-Line Gas/Liquid Filter |