missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE Industrial Tank

Kartell™ HDPE Industrial Tank is useful in everyday laboratory practice and features a comfortable handle with a secure seal-cap following DIN recommendations.

Brand:  Kartell™ 0178000

Code : A0

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10335443

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

HDPE 2L GL45
Specifications

Specifications

Seal-cap
HDPE
175 mm
115 mm
GL45
2 L
155 mm
24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers