missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection

Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.

Brand:  Mettler Toledo™ 30281912

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15549651

  • 163.50 € / Each


Estimated Shipment
07-02-2020
Add to basket

Description

Description

  • Detachable
  • Resuable
Specifications

Specifications

MultiPin-Lemo 00/2 x 4mm banana
1.2m
Mettler Toledo InLab sensors
To connect Mettler Toledo InLab sensors to any meter
Videos
Documents