Learn More
Cytiva Inlet Filter Kit
Filters for buffer inlets as used with Inlet filter holder kit. Cytiva Inlet Filter Kit is a set of 5 polypropylene filters with 5 support screens.
Brand: Cytiva 11-0004-14
Code : GE
Additional Details : Weight : 0.00180kg
Description
This LC (liquid chromatography) accessory consists of five polypropylene filters and five support screens for buffer inlets, as used with an inlet filter holder kit.
- Outer diameter: 15mm
- Wetted parts resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane and 100% Tetrahydrofuran
Specifications
Liquid Chromatography of Biomolecules | |
Polypropylene | |
15 mm |
Inlet Filter Holder | |
5 Filters and 5 Support Screens |