Learn More
Eppendorf™ Innova™ Benchtop Open Air Shaker
Brand: Eppendorf™ M1190-0003
Code : 99
Additional Details : Weight : 0.50000kg
Specifications
19 mm | |
± 1 RPM | |
240 V, 50/60 Hz | |
50/60 Hz | |
37 x 35.5 x 14.6 cm | |
35.5 cm |
15 kg | |
25 to 500 rpm | |
35 VA | |
28 cm | |
14.6 cm | |
2000 |