missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Starsys™ Undercounter Unit

Contains black phenolic island countertop (no backsplash)

Brand:  Metro™ SXRMWS29TS2

Additional Details : Weight : 66.04000kg

Product Code. 15937251

  • 2350.00 € / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

Unit includes:
  • Single-wide non-locking mobile workcenter
  • With 7.62cm (3 in.) diameter total-lock casters
  • Black phenolic island countertop (no backsplash) 2.54cm (1 in.)
  • One polymer drawer 11.4cm (4.5 in.)
  • One polymer drawer 7.62cm (3 in.)
  • One polymer drawer 19.1cm (7.5 in.)
  • One polymer drawer 15.2cm (6 in.)
  • Black drawer pulls
Specifications

Specifications

Metro Starsys Undercounter Unit
633 mm
732 mm
577 mm
24.9 in.
28.8 in.
22.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers