missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrightStar™-Plus Positively Charged Nylon Membrane, 30 cm x 45 cm

For use with radiolabeled and nonisotopic probes for northern and other blot applications

Brand:  Invitrogen™ AM10102

Additional Details : Weight : 0.36000kg

Product Code. 10544045

  • 248.00 € / Each
In Stock
to see stock.
Add to basket

Description

Description

  • For consistent, reliable results with minimal background
  • One roll of 30 x 45cm is provided
  • This membrane exhibits exceptionally low nonspecific binding in nonisotopic detection applications when used with proprietary Locking and wash solutions

DNA and RNA Purification and Analysis, Dot Blotting, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, RNA Extraction, Southern Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
Nylon Membrane
Nylon Membrane
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.