missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IP-10 (CXCL10) Human ProQuantum Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Brand: Invitrogen™ A35578
498.69 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.22720kg
Description
The chemokine IP-10 (or CXCL10) is a chemokine made by monocytes,endothelial cells and fibroblasts in response to treatment with IFNgamma. IP-10 functions as a chemoattractant to cells expressing the G protein-coupled receptor,CXCR3,which is found mainly on activated T cells and NK cells. IP-10 plays an important role in Th1 type inflammatory diseases and autoimmune diseases such as,Hashimoto′s thyroiditis,Graves′ disease and Type 1 diabetes mellitus.Specifications
P02778 | |
<0.05pg/mL | |
ProQuantum | |
Human | |
qPCR Instruments | |
C7; IFI10; INP10; IP-10; SCYB10; crg-2; gIP-10; mob-1 | |
FAM™ | |
RUO | |
-20°C | |
30 min. |
0.064 – 10000pg/mL | |
Taqman™ Real-time PCR | |
Plasma, Serum, Supernatant | |
Cytokine protein quantitation | |
3627 | |
Assay Dilution Buffer<Br>Antibody-Conjugate A<Br>Antibody-Conjugate B<Br>Antibody-Conjugate Dilution Buffer<Br>Standard<Br>Master Mix<Br>Ligase | |
96 Tests | |
Serum, 2-5 μL; Supernatant, 2-5 μL; Plasma, 2-5 μL | |
Human | |
2 hr. |