missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic Wash Buffer I

PureLink Genomic DNA Mini Kit and PureLink 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in a familiar silica spin-column or semi-skirted 96-well plate format.

Brand:  Invitrogen™ K182303

Additional Details : Weight : 0.50000kg

Product Code. 10791764

  • 130.00 € / Each
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: PureLink Genomic Spin Columns, Collection Tubes, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual. The PureLink 96 Genomic DNA Kit includes PureLink Genomic Binding Plates, Wash Plates, Deep Well Plates, Foil Tape, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual.

Description

Description

PureLink Genomic DNA Mini Kit and PureLink 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in a familiar silica spin-column or semi-skirted 96-well plate format.

  • Obtain higher gDNA yields and purity—optimized spin-column membrane and buffer formulations
  • Save time and money—use with large range of sample sizes (e.g., 100 μL to 1 mL blood); 96-well version does not require special equipment
  • Increase flexibility—one kit works with variety of sample types

How it works

  • PureLink Genomic DNA Mini Kit protocol improves upon easy, familiar spin-column methods currently in use
  • Columns are newly configured to bind and release DNA more efficiently
  • Samples are lysed in optimized buffer formulated to enhance proteinase K activity and eliminate protein contamination
  • Powerful wash buffers remove all traces of protein and salt
  • DNA is eluted in low-salt buffer to allow for pH stabilization of the DNA in storage

For higher throughput, use the PureLink 96 Genomic DNA Kit

  • In this 96-well version, sample processing is performed with either vacuum manifold or centrifugation at only 2,250 g, allowing more flexibility in centrifuge choice
  • The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations

Higher yields and purity

  • With the PureLink Genomic DNA Kits, get greater yields of higher-purity gDNA (determined by A260/A280 measurements) with all sample types - from bacteria to tissues to blood to cells
  • PureLink 96 Genomic DNA Kit outperforms competitor products in both DNA yield and purity for multiple sample types, including blood
  • A260/A280 and A260/A230 ratios of PureLink gDNA are higher than those obtained with other kits, indicating no contamination carryover of protein or guanidine using the enhanced PureLink Lysis and Wash Buffers

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Wash Buffer
Store at room temperature.
Bacteria, Blood, Blood Spot, Cells, Swab, Tissue
Genomic Wash Buffer I
Room Temperature
Spin Column
Kit
PureLink™
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.