Learn More
Invitrogen™ UltraPure™ Herring Sperm DNA Solution
Blocks non-specific attachment of probe to surface of membrane
Brand: Invitrogen™ 15634017
Additional Details : Weight : 0.02000kg
Description
UltraPure DNA solutions from Calf Thymus, Herring Sperm, or Salmon Sperm are ready-to-use, sheared DNA solutions that are used directly in the preparation of prehybridization and hybridization solutions. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free and RNase-free (DEPC-treated) distilled, deionized water and are sheared to an average size of ≤2,000 bp. The concentration is adjusted to 10 mg/mL.
Order Info
Shipping Conditions: Dry Ice
Specifications
Store at -20°C. | |
Block Non-Specific Hybridization | |
Herring Sperm DNA Solution | |
Herring Sperm DNA |
Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting | |
UltraPure™ | |
5 x 1 mL | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.