Learn More
Iodine-Bromide solution EU Pharmacopoeia, Fisher Chemical™
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Brand: Fisher Chemical RP 107 1000
Code : 04
Additional Details : Weight : 1.00000kg Transport : UN number : 2789 Chem class : 8 Pack group : II
Specifications
0.02365M (0.0473N) | |
1000 mL | |
Eu. Ph. |
Glass Bottle | |
EU Ph Ref: 1045901 | |
Iodine bromide solution. |
Safety and Handling
- Iodine bromide solution. European Pharmacopoeia Ref: 1045901 (RP 107)
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .