missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Puriss., ≥99.5% (RT), particles (round), Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 184°C (literature) |
Melting Point | 113°C (literature) |
CAS Max % | 100.0000% |
Quantity | 1 kg |
Linear Formula | I2 |
UN Number | UN3495 |
Beilstein | 3587194 |
IUPAC Name | diiodine |
Formula Weight | 253.81 |
Percent Purity | ≥99.5% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'