missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 99%, stabilized, ACROS Organics™

Brand:  Acros Organics 122370025

Code : 29

Additional Details : CAS Number : 74-88-4 Weight : 4.00000kg Transport : UN number : 2644 Chem class : 6.1 Pack group : I

Packaging Glass bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11934501

  • 547.00 € / 2.50kg

Estimated Shipment: 15-07-2021

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

Alkylation reagent
Specifications

Chemical Identifiers

74-88-4
141.94
INQOMBQAUSQDDS-UHFFFAOYSA-N
6328
iodomethane
CH3I
MFCD00001073
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
CHEBI:39282
CI

Specifications

Iodomethane
Authentic
1.5290 to 1.5310
Copper
0.1% max.
Undesignated
2.5kg
98.5% min. (GC)
CH3I
01, 69
15, 6159
Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide
CI
141.94
CHEBI:39282
Liquid
2.2800g/mL
Glass bottle
2.28
0.5 mPa.s (20°C)
42.5°C
-66.0°C
74-88-4,7440-50-8
CH3I
MFCD00001073
01,682; 02,274; 03,202; 04,341; 05,447; 06,384; 08,342; 09,308; 15,118
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
INQOMBQAUSQDDS-UHFFFAOYSA-N
iodomethane
6328
141.94
99%
Videos
SDS Documents
Certificates