Learn More
Iodosobenzene diacetate, 98+%, Thermo Scientific Chemicals
Description
Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 159°C to 161°C |
Quantity | 25 g |
Beilstein | 1879369 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 322.1 |
Percent Purity | ≥98% |
Chemical Name or Material | Iodosobenzene diacetate |
RUO – Research Use Only