missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride, Thermo Scientific™

Topoisomerase I inhibitor

166.00 € - 514.00 €

Chemical Identifiers

CAS 100286-90-6
Molecular Formula C33H39ClN4O6
Molecular Weight (g/mol) 623.15
MDL Number MFCD01862255
InChI Key GURKHSYORGJETM-WAQYZQTGSA-N
Synonym irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
PubChem CID 74990
ChEBI CHEBI:5971
IUPAC Name hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
SMILES [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473449
View Documents
Thermo Scientific Alfa Aesar
J60743.MF
50mg
166.00 €
50mg
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
 
 Restricted Item
15463449
View Documents
Thermo Scientific Alfa Aesar
J60743.MD
250mg
514.00 €
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 25-08-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

100286-90-6
623.15
GURKHSYORGJETM-WAQYZQTGSA-N
74990
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
C33H39ClN4O6
MFCD01862255
irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
CHEBI:5971
[H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O

Specifications

100286-90-6
MFCD01862255
irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
GURKHSYORGJETM-WAQYZQTGSA-N
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
74990
623.14
C33H39ClN4O6
14,5091
Soluble in DMSO or DMF at approximately 20mg/ml; Sparingly soluble in aqueous buffers; /n
[H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
623.15
CHEBI:5971
Irinotecan hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'rtecsNumber' : DW1061000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

Product Identifier
  • Irinotecan hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Irinotecan hydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.