missing translation for 'onlineSavingsMsg'
Learn More

Iron, 99%, powder, -70 mesh (<212 micron), Thermo Scientific Chemicals

Catalog Number p-3734584
15.98 EUR valid until 2024-07-31
Use promo code "23251" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Color Gray
Melting Point 1535.0°C
Boiling Point 3000.0°C
Density 7.8600g/mL
Assay Percent Range 99%
Quantity 100 g
Fieser 01,519; 02,229; 03,167; 05,357; 09,257; 14,184; 16,189; 17,157
Merck Index 15, 5137
Formula Weight 55.84
Percent Purity 99%
Packaging Plastic bottle
Form Fine Powder
Chemical Name or Material Iron
Show More Show Less
compliance-icons
Product Identifier
  • Iron, powder
Signal Word
  • Danger
Hazard Category
  • Self-heating substance Category 1
Hazard Statement
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410-Keep cool. Protect from sunlight.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'