missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) chloride hexahydrate, 99+%, extra pure, Thermo Scientific Chemicals
328.68 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Boiling Point | 280.0°C to 285.0°C |
Melting Point | 37.0°C |
Color | Yellow to Brown |
Physical Form | Lump |
Quantity | 5 kg |
Assay Percent Range | 99% min. (Iodometry) |
Linear Formula | FeCl3·6H2O |
Fieser | 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158 |
Merck Index | 15,4046 |
Solubility Information | Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'