missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, 99+%, extra pure, ACROS Organics™

Brand:  Acros Organics 125030025

Code : 29

Additional Details : CAS Number : 10025-77-1 Weight : 2.55000kg Transport : UN number : 3260 Chem class : 8 Pack group : III

Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11954181

  • 239.00 € / 2.50kg

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lewis acid
Specifications

Chemical Identifiers

10025-77-1
Cl3Fe·6H2O
270.29
Ferric chloride hexahydrate

Specifications

Iron(III) chloride hexahydrate
99% min. (Iodometry)
FeCl3·6H2O
15,4046
Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol,acetone and ether
270.29
99+%
500ppm max.
Plastic bottle
1500ppm max
Yellow to Brown
2.5kg
10025-77-1
Cl3Fe·6H2O
01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Ferric chloride hexahydrate
270.29
Lumps
Extra Pure
0.05% max. (10 g in 50 ml H2O:HCl (49:1))
Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether
280.0°C to 285.0°C
37.0°C
Videos
SDS Documents
Certificates